Human recombinant Hsp47 H Rec
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Produced in E. coli.
Specifications
- Protein/peptide type:Recombinant
- Species:Human
- Protein/peptide name:Hsp47 H Rec
- Purity:>90% (SDS-PAGE)
- Molecular weight:~47 kDa
- Formulation:Liquid, in 30 mM TRIS, pH 8,0, containing 100 mM sodium chloride, 1 mM DTT, and 0,4 M arginine
- Purification:Purified by multi-step chromatography
Specifications
Frequently Bought Together