Adenovirus 5 Hexon
Catalog # ENZOALX6301200100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 µG
- Seq:02
- Protein/peptide name:Adenovirus 5 Hexon
- Purity:>95% (SDS-PAGE)
- Formulation:Liquid, 10 mM BIS-TRIS propane and 375 mM sodium chloride
- Purification:Purified protein.
Specifications
About this item
From adenovirus type 5.
Native protein.