Order Entry
Austria
ContactUsLinkComponent
Human recombinant NF-kappaB p50 subunit (35-381) (GST-tag) (from E. coli), GST Tag
Human recombinant NF-kappaB p50 subunit (35-381) (GST-tag) (from E. coli), GST Tag
Catalog # ENZOBMLUW99800050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant NF-kappaB p50 subunit (35-381) (GST-tag) (from E. coli), GST Tag
Catalog # ENZOBMLUW99800050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    GST Tag
  • Protein/peptide name:
    NF-kappaB p50 subunit (35-381) (GST-tag)

Specifications

About this item

Produced in E. coli.