Human recombinant IkappaBalpha (from E. coli)
Catalog # ENZOBMLUW99750050
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:IkappaBalpha
Specifications
About this item
Produced in E. coli.