Human recombinant BAP1 (from E. coli), HIS Tag
ENZOBMLUW98550050
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Tag sequence:HIS Tag
- Protein/peptide name:BAP1
Produced in E. coli. Human BAP1 is fused at the N-terminus to a His-tag.