Di-Ubiquitin (K48-linked)
Catalog # ENZOBMLUW98000100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 µG
- Seq:02
- Protein/peptide name:Di-Ubiquitin (K48-linked)
- Purity:>95% (SDS-PAGE)
- Formulation:Liquid, containing 50 mM TRIS pH 7,5, 1 mM EDTA and 5 mM DTT.
- Purification:Ubiquitin mutants [K48C] and [D77] were expressed in BL21 (λDE3) E. coli. Proteins were purified by cation exchange chromatography. Di-ubiquitin was synthesised by linking [K48C] ubiquitin and [D77] ubiquitin in the presence of ubiquitin conjugating enzyme E2-25K (UbcH1) (Prod. No. BML-UW9020) and human ubiquitin activating enzyme E1 (Prod. No. BML-UW9410). The product was purified by ion exchange chromatography. Purity was confirmed by SDS-PAGE and protein concentration estimated by absorbance at 280 nm.
Specifications
About this item
Produced in E. coli and biochemically synthesized in vitro.