Human recombinant UCH-L3 (from E. coli), HIS tag
Catalog # ENZOBMLUW97450050
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Tag sequence:HIS Tag
- Protein/peptide name:UCH-L3
Specifications
About this item
Produced in E. coli.