Order Entry
Austria
ContactUsLinkComponent
Human recombinant UCH-L1 (from E. coli), HIS tag
Human recombinant UCH-L1 (from E. coli), HIS tag
Catalog # ENZOBMLUW97400050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant UCH-L1 (from E. coli), HIS tag
Catalog # ENZOBMLUW97400050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    HIS Tag
  • Protein/peptide name:
    UCH-L1

Specifications

About this item

Produced in E. coli.