Human recombinant pro-ISG15 (from E. coli), HIS Tag
Catalog # ENZOBMLUW92300500
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:500 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Tag sequence:HIS Tag
- Protein/peptide name:pro-ISG15
- Purity:>95% (SDS-PAGE)
- Formulation:Liquid. In 20 mM HEPES, pH 8,0, containing 50 mM sodium chloride and 1 mM DTT.
Specifications
About this item
Produced in E. coli. Human pro-ISG15 is fused at the N-terminus to a His-tag.
ISG15 is expressed in its pro-form with a short C-terminal extension, which is processed to expose the C-terminal glycine residue that is essential for conjugation to target proteins.