Order Entry
Austria
ContactUsLinkComponent
Human recombinant pro-SUMO-1 (from E. coli), HIS Tag
Human recombinant pro-SUMO-1 (from E. coli), HIS Tag
Catalog # ENZOBMLUW91900500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant pro-SUMO-1 (from E. coli), HIS Tag
Catalog # ENZOBMLUW91900500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    500 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    HIS Tag
  • Protein/peptide name:
    pro-SUMO-1

Specifications

About this item

Produced in E. coli. Human pro-SUMO-1 is fused at the N-terminus to a His-tag.