Ubiquitin vinyl-methylester (HA tag), HA tag
ENZOBMLUW08800025
:
- Pk:25 µG
- Seq:02
- Tag sequence:HA Tag
- Reconstitution instructions:Reconstitute with aqueous buffer up to 5 mg/ml (~500 μM)
- Protein/peptide name:Ubiquitin Vinyl-methylester (HA tag)
- Purity:>95% (HPLC)
- Formulation:Lyophilised
Synthetic