Order Entry
Austria
ContactUsLinkComponent
Human recombinant PRL-1 (from E. coli), HIS Tag
Human recombinant PRL-1 (from E. coli), HIS Tag
Catalog # ENZOBMLSE5560050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant PRL-1 (from E. coli), HIS Tag
Catalog # ENZOBMLSE5560050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    HIS Tag
  • Protein/peptide name:
    PRL-1
  • Purity:
    >90% (SDS-PAGE)
  • Concentration:
    0,05 μg/μl
  • Formulation:
    Liquid. In 25 mM TRIS-HCl, pH 8,0, containing 100 mM sodium chloride, 0,05% Tween 20, 20% glycerol and 3 mM dithiothreitol.

Specifications

About this item

Produced in E. coli. The catalytic domain of PRL-1 (aa 2-173) is fused at the N-terminus to a His-tag.