Human recombinant PDE4B2 (catalytic domain) (from E. coli)
ENZOBMLSE5220020
:
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:PDE4B2 (catalytic domain)
Produced in E. coli. Active catalytic domain of phosphodiesterase 4B2.