Mouse MMP2 (from fibroblasts)
Catalog # ENZOBMLSE5050005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:5 µG
- Source:Fibroblasts
- Species:Mouse
- Seq:02
- Protein/peptide name:MMP2
- Purification:Free of TIMPs and other MMPs.
Specifications
About this item
Isolated from mouse fibroblasts. Primarily latent form.