Human recombinant HDAC2 (1-488), HIS tag
Catalog # ENZOBMLSE5000050
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Tag sequence:HIS Tag
- Protein/peptide name:HDAC2 (1-488)
Specifications
About this item
Highly active
Produced in insect cells.