Order Entry
Austria
ContactUsLinkComponent
Human recombinant c-erbB-2
Human recombinant c-erbB-2
Catalog # ENZOBMLSE4600010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant c-erbB-2
Catalog # ENZOBMLSE4600010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    c-erbB-2

Specifications

About this item

Produced in insect cells. Active c-erbB2 (aa 679-1255).