Order Entry
Austria
ContactUsLinkComponent
Human recombinant MKP-3 (from insect cells (Sf9))
Human recombinant MKP-3 (from insect cells (Sf9))
Catalog # ENZOBMLSE4540100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant MKP-3 (from insect cells (Sf9))
Catalog # ENZOBMLSE4540100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells (Sf9)
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    MKP-3

Specifications

About this item

Produced in Sf9 cells. Recombinant full length MKP-3.