Order Entry
Austria
ContactUsLinkComponent
 
Human recombinant Flt1,HIS Tag
  ENZOBMLSE4450010
 :  
Human recombinant Flt1,HIS Tag
  ENZOBMLSE4450010
 :  

 

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    HIS Tag
  • Protein/peptide name:
    Flt1

 

 

Produced in insect cells. Active Flt1 (aa 783- end).