Order Entry
Austria
ContactUsLinkComponent
Human recombinant DPPIV
Human recombinant DPPIV
Catalog # ENZOBMLSE4340025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant DPPIV
Catalog # ENZOBMLSE4340025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 Mu
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    DPPIV

Specifications

About this item

Highly active

Produced in insect cells. Active recombinant human DPPIV (aa 29-766).This represents a naturally-occurring cleaved (soluble) form of DPPIV.