Human recombinant CDC25B (from E. coli)
Catalog # ENZOBMLSE3650050
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:CDC25B
Specifications
About this item
Produced in E. coli. Full length human CDC25B.