Order Entry
Austria
ContactUsLinkComponent
Human native factor Xa (from plasma)
Human native factor Xa (from plasma)
  ENZOBMLSE3620100
 :  
Human native factor Xa (from plasma)
  ENZOBMLSE3620100
 :  

 

  • Pk:
    100 µG
  • Protein/peptide type:
    Native
  • Source:
    Plasma
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    Factor Xa
  • Grade:
    purified

 

 

Isolated from human plasma.