Human recombinant MMP9 (catalytic domain) (from E. coli)
Catalog # ENZOBMLSE3600010
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:MMP9 (catalytic domain)
- Purity:>95% (SDS-PAGE)
- Concentration:0,5 µg/µl
- Formulation:Liquid. In 50 mM TRIS, pH 7,5, containing 5 mM calcium chloride, 300 mM sodium chloride, 20 µM zinc chloride, 0,5% Brij-35 and 30% glycerol.
Specifications
About this item
Produced in E. coli. Active recombinant matrix metalloproteinase-9 (MMP-9, gelatinase B, 92 kDa type IV collagenase) cloned from human cDNA.
- Activity toward its targets, such as gelatin, casein, or peptide substrates, is unaffected
The enzyme consists of residues Phe107-Pro449 (NM_004994), which comprises the catalytic/fibronectin domain of human MMP-9, with a C-terminal purification tag. This represents a naturally-occurring active form of MMP-9 which lacks the C-terminal hemopexin domain.