Order Entry
Austria
ContactUsLinkComponent
Human recombinant PKCμ (GST-tag) (from insect cells (Sf9)), GST Tag
Human recombinant PKCμ (GST-tag) (from insect cells (Sf9)), GST Tag
Catalog # ENZOBMLSE3480005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant PKCμ (GST-tag) (from insect cells (Sf9)), GST Tag
Catalog # ENZOBMLSE3480005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells (Sf9)
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    GST Tag
  • Protein/peptide name:
    PKCμ (GST-tag)

Specifications

About this item

Produced in Sf9 cells. Active, full length protein kinase C mu (PKCµ, protein kinase D, PKD) is fused at the N-terminus to a GST-tag.