Human Recombinant 14-3-3gamma (GST-tag) (from E. coli), GST Tag
Catalog # ENZOBMLSE3130100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Tag sequence:GST Tag
- Protein/peptide name:14-3 -3 gamma (GST-tag)
Specifications
About this item
Produced in E. coli. Human 14-3-3gamma is fused at the N-terminus to a GST-tag.