Human recombinant TNF alpha (2)
ENZOBMLSE3030100
:
- Pk:100 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Protein/peptide name:TNF alpha (2)
- Formulation:Sterile, lyophilised powder.
Produced in E. coli as a single, non-glycosylated polypeptide.