Order Entry
Austria
ContactUsLinkComponent
Human recombinant CDK1
Human recombinant CDK1
Catalog # ENZOBMLSE2950010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant CDK1
Catalog # ENZOBMLSE2950010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Protein synonyms:
    Cyclin B
  • Protein/peptide name:
    CDK1

Specifications

About this item