Order Entry
Austria
ContactUsLinkComponent
Human recombinant MMP-14 (catalytic domain) (112-298)
Human recombinant MMP-14 (catalytic domain) (112-298)
Catalog # ENZOBMLSE2590010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant MMP-14 (catalytic domain) (112-298)
Catalog # ENZOBMLSE2590010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    MMP-14 (catalytic domain) (112-298)

Specifications

About this item

Produced in E.coli. Active Matrix Metalloproteinase-14 (MMP-14, Membrane-Type Matrix Metalloproteinase 1, MT1-MMP) catalytic domain from human cDNA.

The enzyme consists of the catalytic domain of human MMP-14 (Tyr112-Arg298, NM_004995) with a C-terminal purification tag. This represents a naturally-occurring active form of MMP-14 which lacks the C-terminal hemopexin domain.

MMPs lacking this domain cannot cleave native collagens; however, activity toward other targets such as gelatin, casein, or peptide substrates is unaffected.