Order Entry
Austria
ContactUsLinkComponent
Human recombinant MMP8 (catalytic domain) (from E. coli)
Human recombinant MMP8 (catalytic domain) (from E. coli)
Catalog # ENZOBMLSE2550010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant MMP8 (catalytic domain) (from E. coli)
Catalog # ENZOBMLSE2550010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    MMP8 (catalytic domain)

Specifications

About this item

Produced in E. coli. Active Matrix Metalloproteinase-8 (MMP-8, neutrophil collagenase, collagenase-2) catalytic domain from human cDNA.

The enzyme consists of the catalytic domain of human MMP-8 (Phe99-Gln269, NM_002424) with a C-terminal purification tag. This represents a naturally-occurring active form of MMP-8 which lacks the C-terminal hemopexin domain1.

MMPs lacking this domain cannot cleave native collagens; however, activity toward other targets such as gelatin, casein, or peptide substrates is unaffected.