Order Entry
Austria
ContactUsLinkComponent
Human recombinant SIRT1 (from E. coli), His-tag
Human recombinant SIRT1 (from E. coli), His-tag
Catalog # ENZOBMLSE2390100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant SIRT1 (from E. coli), His-tag
Catalog # ENZOBMLSE2390100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 Vial
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    HIS Tag
  • Protein/peptide name:
    SIRT1
  • Purity:
    >50% (SDS-PAGE)

Specifications

About this item

Highly active

Produced in E. coli. Contains a N-terminal His-tag.

This protein is produced from human cDNA, expressed in E. coli. Sequence identical to that at Genbank accession #NM012238