Order Entry
Austria
ContactUsLinkComponent
Human recombinant Granzyme B (from yeast)
Human recombinant Granzyme B (from yeast)
Catalog # ENZOBMLSE2385000
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant Granzyme B (from yeast)
Catalog # ENZOBMLSE2385000
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 Vial
  • Protein/peptide type:
    Recombinant
  • Source:
    Yeast
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    Granzyme B

Specifications

About this item

Produced in yeast. Recombinant human granzyme B (aa 21-247).