Human recombinant MMP2 (catalytic domain)
ENZOBMLSE2370010
:
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Protein/peptide name:MMP2 (catalytic domain)
- Formulation:In TRIS-HCl, pH 7,5, containing 300 mM NaCl, 5 mM CaCl2, 20 µM ZnCl2 , 0,5% Brij-35 and 20% Glycerol.
Produced in yeast. Active recombinant matrix metalloproteinase-2 (MMP-2, gelatinase A, 72kDa type IV collagenase) cloned from human cDNA.
The enzyme consists of residues Tyr110-Asp452 (NM_004530), which comprises the catalytic/fibronectin domain of human MMP-2, with a C-terminal purification tag. This represents a naturally occurring active form of MMP-2 which lacks the C-terminal hemopexin domain.
MMPs lacking this domain cannot cleave native collagens; however, activity toward other targets such as gelatin, casein, or peptide substrates is unaffected.