Order Entry
Austria
ContactUsLinkComponent
Human recombinant Leptin (from E. coli)
Human recombinant Leptin (from E. coli)
Supplier:  ENZO LIFE SCIENCES
Certificates
Human recombinant Leptin (from E. coli)
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein/peptide name:
    Leptin
  • Purity:
    >95%
  • Purification:
    Highly purified and biologically active.
  • Formulation:
    Lyophilised

Specifications

Frequently Bought Together