Z-Leu-Arg-Gly-Gly-Amc
Catalog # ENZOBMLP8010005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:5 mg
- Seq:02
- Protein/peptide name:Z-Leu-Arg-Gly-Gly-AMC
- Purity:95%
Specifications
About this item
Substrate for ubiquitin C-terminal hydrolases (UCHs)
Fluorogenic substrate for ubiquitin C-terminal hydrolases (UCHs), including UCH-L3 and Isopeptidase T. Sequence is based on the C-terminal residues of ubiquitin. Ex.: 380 nm Em.: 460 nm.