IKK-NBD control peptide
Catalog # ENZOBMLP6080500
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:0,5 mg
- Seq:02
- Protein/peptide name:IKK-NBD control Peptide
- Purity:95%
- Molecular weight:3463,0
- Tested applications:Control peptide
Specifications
About this item
Negative control for IKK-NBD peptide
Negative control peptide for IKK-NBD peptide. Sold under license from Vanderbilt University