YEEI Peptide (Biotin)
Catalog # ENZOBMLP3020200
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:200 µG
- Conjugation:Biotin
- Seq:02
- Protein/peptide name:YEEI Peptide
- Purity:>97%
Specifications
About this item
Binds and activates src family kinases
This biotinylated probe binds to SH2 domains of Src family members including Src, Lck, and Fyn.