IGF 1Rtide
Catalog # ENZOBMLP2570001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:02
- Protein/peptide name:IGF 1R tide
- Purity:95%
Specifications
About this item
Protein kinase substrate
This peptide, KKKSPGEYVNIEFG, is derived from human IRS-1 and is a substrate for TRK1, JAK2, and RET kinases.