Order Entry
Austria
ContactUsLinkComponent
HER2 Substrate
HER2 Substrate
Catalog # ENZOBMLP2420001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
HER2 Substrate
Catalog # ENZOBMLP2420001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    02
  • Protein/peptide name:
    HER2 substrate
  • Purity:
    95%

Specifications

About this item

This peptide, GGMEDIYFEFMGGKKK, identified by screening a degenerate peptide library, is a substrate for several kinases, including HER-2/c-neu/erb-B-2, Tie-2, cFMS, and cKit.