SAMS Peptide
Catalog # ENZOBMLP2310001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:02
- Protein/peptide name:SAMS Peptide
- Purity:95%
Specifications
About this item
AMPK substrate
This peptide, HMRSAMSGLHLVKRR, is derived from rat acetyl-CoA carboxylase and is known to be a good substrate for AMPK.