ContactUsLinkComponent
[pThr183, pTyr185] MAP kinase (177-189)
[pThr183, pTyr185] MAP kinase (177-189)
Catalog # ENZOBMLP1640001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
[pThr183, pTyr185] MAP kinase (177-189)
Catalog # ENZOBMLP1640001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Pk:
    1 mg
  • Seq:
    02
  • Protein/peptide name:
    [pThr183, pTyr185] MAP kinase (177-189)
Specifications
About this item
VHR substrate
Diphosphopeptide with sequence, DHTGFLpTEpYVATR, corresponding to MAP kinase residues 177-189 inclusive of putative activation sites. Peptide can be used as a VHR phosphatase substrate (BML-SE333). Phosphate release may be quantified using BML-AK111 malachite green colorimetric reagent. Phosphate release is much more rapid for the phosphotyrosine residue compared to the phosphothreonine residue, both in vitro and in vivo.