Order Entry
Austria
ContactUsLinkComponent
PKCzeta Substrate
PKCzeta Substrate
Catalog # ENZOBMLP1560001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
PKCzeta Substrate
Catalog # ENZOBMLP1560001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    02
  • Protein/peptide name:
    PKCzeta substrate

Specifications

About this item

This peptide sequence, RRFKRQGSFFYFF, was selected from a peptide library for optimal phosphorylation by protein kinase C zeta (PKCζ).