ContactUsLinkComponent
Z-GPR-Amc
Z-GPR-Amc
Catalog # ENZOBMLP1420020
Supplier:  Enzo Life Sciences
CAS Number:  
Z-GPR-Amc
Catalog # ENZOBMLP1420020
Supplier:  Enzo Life Sciences
CAS Number:  
Specifications
  • Pk:
    20 mg
  • Seq:
    02
  • Protein/peptide name:
    Z-GPR-AMC
Specifications
About this item
Cathepsin K substrate
Fluorogenic substrate specific for cathepsin K, among other cathepsin family members. This substrate is also cleaved by granzyme A, thrombin, tryptase, trypsin, and kallikreins. Ex.: 365 nm, Em.: 440 nm, although the following Ex/Em can also be used: 355,380/450,460. This substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144).