CSH103 substrate
Catalog # ENZOBMLP1040500
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:500 µG
- Seq:02
- Protein/peptide name:CSH103 Substrate
Specifications
About this item
CDK substrate
This sequence, ADAQHATPPKKKRKVED, is SV40 L Tag (118-138) with Ser to Ala120 and Ala123 substitutions to increase specificity. Substrate is specific for p34cdc2 (CDK1) and is a poor substrate for CKI, II, PKA, GSK3, PKC, CaMKII, and phosphorylase b kinase. Can be used in cell lysates using TCA/phosphocellulose assay. Km=74 µM for active complex with cyclin B.