Ac-Arg-Leu-Arg-AMC
Catalog # ENZOBMLAW97850005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:5 mg
- Seq:02
- Protein/peptide name:Ac-Arg-Leu-Arg-AMC
- Purity:>97% (HPLC)
Specifications
About this item
This fluorogenic peptide is often the substrate of choice for assaying the trypsin-like activity of purified proteasomes. Benefits include a low Km, high specific activity, and the fact that the substrate is cleaved exclusively at the R-AMC bond.