alpha-Latrotoxin
ENZOALX630027C040
New Product
:
- Pk:40 µG
- Source:
- Species:
- Seq:02
- Environmentally Preferable:
- Reconstitution instructions:Reconstitute in 0,5 ml of distilled cold water. Stir very gently. Do not shake or vortex. Add 0,5 ml glycerol. Stock solution received contains 50% glycerol and 300 nM α-latrotoxin.
- Protein/peptide name:alpha-Latrotoxin
- Purity:>97%
- Formulation:Lyophilised
Neurotoxin
- Isolated from Latrodectus tredecimguttatus
Causes massive neurotransmitter release from a wide variety of central and peripheral synaptic junctions of vertebrates using Ca²⁺-dependent and Ca²⁺-independent pathways. A useful pharmacological tool in the studies of synaptic vesicles exocytosis of different neutrotransmitters.
: Quantity: Determined by Lowry and Pierce method.