omega-Agatoxin IVA
ENZOALX630001C100
New Product
:
- Pk:100 µG
- Source:
- Species:
- Seq:02
- Environmentally Preferable:
- Protein/peptide name:Omega-Agatoxin IVA
- Purity:>98%
Ca²⁺ channel blocker
Selective P-type Ca²⁺ channel blocker. Fast acting and potent neurotoxin.