Order Entry
Austria
ContactUsLinkComponent
 
Human;Mouse recombinant Fc(Human):RANKL (soluble)
  ENZOALX522131C010
 :  
Human;Mouse recombinant Fc(Human):RANKL (soluble)
  ENZOALX522131C010
 :  

 

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
    Mouse
  • Seq:
    02
  • Reconstitution instructions:
    Reconstitute with 100 µl sterile water.
  • Protein/peptide name:
    Fc(human):RANKL (soluble)
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    0,1 mg/ml after reconstitution
  • Formulation:
    Lyophilised. Contains PBS.

 

 

Produced in HEK 293 cells. The extracellular domain of mouse RANKL (aa 157-316) is fused to the Fc portion of human IgG1.