Order Entry
Austria
ContactUsLinkComponent
Human Recombinant CD137L (soluble)
Human Recombinant CD137L (soluble)
Supplier:  ENZO LIFE SCIENCES
Certificates
Human Recombinant CD137L (soluble)
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    CD137L (soluble)
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    0,1 mg/ml after reconstitution
  • Formulation:
    Lyophilised. Contains PBS and 5% trehalose.
  • Reconstitution instructions:
    Reconstitute with 100 µl sterile distilled water. Further dilutions should be made with medium containing 5% fetal calf serum.

Specifications

Frequently Bought Together