Human recombinant Adiponectin
Catalog # ENZOALX522063C050
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Biological activity:Activates AMPK.
- Reconstitution instructions:Reconstitute with 50 µl sterile distilled water. Further dilutions should be made with medium containing 5% fetal calf serum or other carrier protein.
- Protein synonyms:ACDC
- Protein/peptide name:Adiponectin
- Purity:>90% (SDS-PAGE)
- Concentration:1 mg/ml after reconstitution
- Formulation:Lyophilised. Contains PBS.
Specifications
About this item
Human adiponectin (aa 16-244) is fused at the N-terminus to a linker peptide (14 aa) and a FLAG®-tag.
- Produced in HEK 293 cells
Mimics serum adiponectin by forming high molecular weight (HMW) and hexameric species.