Flagellin
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Isolated from Salmonella typhimurium strain 14028.
Specifications
- Biological activity:Activation of TLR5 in human epithelial cell assays based on NF-κB luciferase fusions, on expression of IL-8 (CXCL8), CCL20, inducible nitric oxide synthase, β-defensin 2 and matrilysin (MMP-7). Maturation of dendritic cells in in vitro and in vivo assays.
- Reconstitution instructions:Reconstitute with 100 µl sterile water.
- Protein/peptide name:Flagellin
- Purity:>95% (SDS-PAGE)
- Concentration:0,1 mg/ml after reconstitution
- Formulation:Lyophilised. Contains PBS.
Specifications
Frequently Bought Together