Order Entry
Austria
ContactUsLinkComponent
 
Microcystis aeruginosa Microcystin LF
 
undefined
Microcystis aeruginosa Microcystin LF

 

  • Species:
    Microcystis aeruginosa
  • Storage conditions:
    –20 °C
  • Protein/peptide name:
    Microcystin LF
  • Purity:
    >95% (HPLC)

 

Frequently Bought Together